Key Documents
Z222488
Keck® Joint Clip
joint size, 35, ball-joint
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
color
orange
General description
Color coded, Delrin® plastic joint clips will not scratch glassware or corrode. Withstand temperatures up to 302 °F (150 °C). Resistant to concentrated alkalis and diluted acids; not resistant to halogen gases or concentrated acids.
Legal Information
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Keck is a registered trademark of Adcuram Lab Technologies GmbH