Z254266
Individual Filter Adapters
size 2
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
Doporučené produkty
size
2
packaging
pkg of 12 ea
H × thickness
21 mm × 2.5 mm
bottom O.D. × I.D.
16 mm × 11 mm
top O.D. × I.D.
27 mm × 22 mm
Související kategorie
General description
For Buchner, conical, fritted-glass, and porcelain filter funnels. Gray neoprene rubber adapters fit singly or can be nested around funnel stem to produce a good fit. Withstands autoclaving.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)