Hlavní dokumenty
Z214825
Sigma-Aldrich
Aldrich® modified Davies condenser
jacket L 400 mm, joint: ST/NS 24/40
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
joint
joint: ST/NS24/40
jacket L
400 mm
General description
Additional internal cooling surface increases efficiency with negligible hold-up when used at a standard 75° angle.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)