Z214396
Ace-Thred™ Thermometer/Tubing Adapter
Twin angled, side joint: ST/NS 24/40, top joint: ST/NS 24/40, bottom joint: ST/NS 24/25, thread I.D. 7.0 mm
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Doporučené produkty
label
D
joint
bottom joint: ST/NS24/25
side joint: ST/NS24/40
top joint: ST/NS24/40
manufacturer/tradename
Ace Glass 526317
thread I.D.
7.0 mm
General description
Threaded nylon bushing with FETFE® O-ring.
Legal Information
Ace-Thred is a trademark of Ace Glass, Inc.
FETFE is a registered trademark of Ace Glass, Inc.
replacement part
Č. produktu
Popis
Stanovení ceny
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)