Key Documents
Z189898
Sigma-Aldrich
Spinplus® magnetic stirring bar
size 1 1/2 in. × 1 1/2 in.
Synonyma:
Stir Bar, Stirbar, Stirring Bar
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
material
PTFE-coated
cross PTFE-coated
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37144-0112
L
38.1 mm
diam.
15.8 mm
size
1 1/2 in. × 1 1/2 in.
General description
Cross-shaped bars create a deep vortex for stable operations.
Legal Information
Spinplus is a registered trademark of SP Scienceware, a division of SP