Hlavní dokumenty
Z177954
Sigma-Aldrich
Scienceware® chemical spoons
L 25 cm
Synonyma:
laboratory spatula, laboratory spoon
About This Item
UNSPSC Code:
41122403
NACRES:
NB.35
material
stainless steel
manufacturer/tradename
Bel-Art H367290025
L
25 cm
spoon end size
1.8 mL , small
5.0 mL , large
General description
Double-ended with large-size spoon on one end and small spoon on the other. Solid, rounded connecting shaft.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)