Z154563
Positioning collar
PTFE/Glass composite, fits shaft o.d., 10 mm
About This Item
UNSPSC Code:
41100000
NACRES:
NB.72
Doporučené produkty
material
PTFE gasket
feature
fits shaft o.d. 10 mm
manufacturer/tradename
Ace Glass 812710
General description
Designed for use with stirring shafts to position shaft in bearing and prevent it from dropping into flask. Gasket prevents scratching bearing and acts as a dust cover.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)