Key Documents
Z125369
Ace-Thred Thermometer/Tubing Adapter
Straight, joint: ST/NS 14/20, thread I.D. 7.0 mm
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
label
A
joint
joint: ST/NS14/20
manufacturer/tradename
Ace Glass 502826
thread I.D.
7.0 mm
General description
Threaded nylon bushing with FETFE® O-ring.
Legal Information
Ace-Thred is a trademark of Ace Glass, Inc.
FETFE is a registered trademark of Ace Glass, Inc.
replacement part