Key Documents
Z120960
Syringe pressure caps
sterile
About This Item
UNSPSC Code:
41121800
NACRES:
NB.03
material
polyolefin
sterility
sterile
packaging
bag of 100 ea
General description
Disposable. For use with Luer lock or regular Luer syring tips, covers entire Luer skirt of syringe.