Hlavní dokumenty
Z118141
Sigma-Aldrich
Straight septum-inlet adapter
inlet I.D. 6 mm, brass
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
material
brass
inlet I.D.
6 mm
General description
1/4 in. NPTM × inlet 6 mm I.D. adapters require septa Z565687 or Z565695.
1/4 in. NPTM × inlet 13.9 mm I.D. adapters require septa Z124591 or Z167282
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)