Key Documents
Z261505
Luer-to-Threaded NPT connector
Micro-Mate® female Luer to 1/8-27 thread, nickel plated
About This Item
UNSPSC Code:
41122000
NACRES:
NB.03
material
nickel plated
manufacturer/tradename
Popper 6415
parameter
45 psi pressure (final assembly water tested)
Související kategorie
Legal Information
Micro-Mate is a registered trademark of Cadence Science