Key Documents
Z116394
Double-ended needle
gauge 12, L 6 in.
About This Item
UNSPSC Code:
41122002
NACRES:
NB.03
packaging
pack of 6 ea
manufacturer/tradename
Cadence Science
L
6 in.
gauge
12
General description
304 SS. One noncoring tip, one flat-cut end.