Key Documents
Z102458
Double-ended needle
gauge 16, L 36 in.
About This Item
UNSPSC Code:
41122002
NACRES:
NB.03
packaging
pack of 1 ea
manufacturer/tradename
Cadence Science
L
36 in.
gauge
16
General description
304 SS. One noncoring tip, one flat-cut end.