Key Documents
SYNF901450G
Synthware pear-shaped Schlenk flask with stopcock
capacity 50 mL, joint: ST/NS 14/20
Synonyma:
Synthware F901450G, modified Schlenk reaction flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
material
borosilicate glass flask
pear-shaped
single-neck
joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F901450G
capacity
50 mL
General description
A suffix of G indicates a glass stopcock, H indicates a high-vacuum valve, no letter suffix indicates a PTFE Stopcock.