Hlavní dokumenty
SYNF780125
Synthware Squibb separatory funnel with glass stopcock
125 mL, stopper size #22
Synonyma:
Synthware F780125, glass separatory funnel
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F780125
capacity
125 mL
125 mL
General description
Supplied complete with a flask length glass stopper of the listed size.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)