Key Documents
SYNA681420
Synthware flushing adapter with PTFE stopcock
joint: ST/NS 14/20
Synonyma:
Synthware A681420, Glass flushing adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A681420
Související kategorie
General description
With 2 mm T-bore PTFE stopcock.