Hlavní dokumenty
DWK11-0051G
Wheaton® Microliter Snap Caps With Septa
green PTFE cap (Push fit)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.11
material
green PTFE cap (Push fit)
feature
cap with septa
packaging
pkg of 100 ea
manufacturer/tradename
WHEATON® 11-0051G
Application
Chromatography
Features and Benefits
  • 11mm Snap Caps with installed septa offered in a wide variety of the most popular configurations.
  • Where noted, the Ring is installed between cap and septa for robotics employing magnetics to move vials.
  • Approved for GC and LC (where noted).
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)