Hlavní dokumenty
BR781525
Brand®1.2 ML Tubes
strip of 8 tubes, polypropylene
Synonyma:
Sample Storage, Tube racks, Tubes
About This Item
UNSPSC Code:
41122804
NACRES:
NB.12
material
polypropylene
sterility
non-sterile
feature
strip of 8 tubes
packaging
pack of 120
manufacturer/tradename
BRAND 781525
capacity
1.2 mL
General description
BIO-CERT® CERTIFIED QUALITY: Free of human DNA, RNase, DNase and Pyrogens
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)