Key Documents
BR122750
BRAND® Media Bottle Cap
polypropylene cap, for 500 mL bottle, pack of 10 ea
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
polypropylene cap
joint
Joints threaded cap (GL56)
feature
for 500 mL bottle
packaging
pack of 10 ea
manufacturer/tradename
BRAND 122750