Key Documents
27519
Supelco
Screw Top Vial, Graduated, Preassembled
volume 15 mL, clear glass vial, thread for 18-400, O.D. × H 21 mm × 70 mm, PTFE/silicone septum
About This Item
UNSPSC Code:
41122001
material
PTFE/silicone septum
black phenolic cap
clear glass
clear glass vial
feature
closure type black phenolic hole cap
closure type screw top vial
packaging
pkg of 100 ea
O.D. × H
21 mm × 70 mm
volume
15 mL
fitting
thread for 18-400
General description
Hole cap is black phenolic.
Septum is blue PTFE on white silicone.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)