Key Documents
RF451817C-EP
Rotor for Eppendorf® 5430/5430R Centrifuges
FA-45-18-17-Cryo, non aerosol tight, to hold, 18 x cryovials, with adapters and lid
About This Item
UNSPSC Code:
41103909
feature
to hold 18 x cryovials
fixed angle rotor
non aerosol tight
with adapters and lid
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022654161 (NA)
Eppendorf® 5427705007 (EU)
parameter
8,324 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5430/5430R
Categorías relacionadas
Legal Information
Eppendorf is a registered trademark of Eppendorf AG