Documentos clave
Z420581
Sigma-Aldrich
Perforated filter plate for Table-Top Buchner funnels
for Z420557
About This Item
Código UNSPSC:
41104925
NACRES:
NB.43
Materiales
high-density polyethylene
esterilidad
non-sterile
envase
pack of 1 ea
fabricante / nombre comercial
Bel-Art H91463-1914
diám. del filtro
36 in.
color
none
Descripción general
Filter large volumes quickly with a polyethylene plastic table-top buchner funnel using your choice of perforated filter plate. Plates are made of 4.8mm (³/₁₆") thick HDPE with 4.8mm (³/₁₆") perforations on 11mm (⁷/₁₆") centers.
Características y beneficios
  • Removable plate for use in precious metals recovery
  • Temperature range maximum 52ºC (125ºF), minimum -20ºC (-4ºF)
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)