Z222712
Drying-tube assembly for Aldrich Chemical Dryer and Chem-Dry apparatus
Complete, joint: ST/NS 34/35
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Productos recomendados
material
colorless tube
glass stopcock
glass tube
round bottom
joint
joint: ST/NS34/35
sterility
non-sterile
feature
graduations: no
binding type
non-treated surface
General description
These replacement drying-tube pieces fit as a replacement parts in the Aldrich Chemical Dryer and Chem-Dry apparatus.
This complete assembly is the drying tube with removable desiccant bulb, 34/35 joint, 3-way glass stopcock. This is for use with the Aldrich Chemical Dryer and Chem-Dry apparatus, Z251984 and Z251992.