Z222399
Metal standard taper clip
joint size, 29
About This Item
Código UNSPSC:
41122413
NACRES:
NB.35
Productos recomendados
Descripción general
Nickel-plated spring-steel joint clips offer increased strength and resistance to heat and cracking over Delrin® plastic type. Securely holds conical joints under pressure at temperatures up to 392 °F (200 °C). Will not scratch glassware.
Información legal
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)