Z150452
Keck® Joint Clip
standard taper, joint size 29, red
About This Item
Código UNSPSC:
41122413
NACRES:
NB.35
Productos recomendados
color
red
Descripción general
Color coded, Delrin® plastic joint clips will not scratch glassware or corrode. Withstand temperatures up to 302 °F (150 °C). Resistant to concentrated alkalis and diluted acids; not resistant to halogen gases or concentrated acids.
Información legal
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Keck is a registered trademark of Adcuram Lab Technologies GmbH
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)