Key Documents
Z150436
Keck® Joint Clip
standard taper, joint size 19, blue
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
color
blue
General description
Color coded, Delrin® plastic joint clips will not scratch glassware or corrode. Withstand temperatures up to 302 °F (150 °C). Resistant to concentrated alkalis and diluted acids; not resistant to halogen gases or concentrated acids.
Legal Information
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Keck is a registered trademark of Adcuram Lab Technologies GmbH
Visite la Librería de documentos
Los clientes también vieron
Slide 1 of 2
1 of 2