Documentos clave
58074
Reservoir for Filtration Apparatus 1
volume 500 mL
About This Item
Código UNSPSC:
41115700
volumen
500 mL
Descripción general
Reservoir for Filtration Apparatus 1 (volume 500 mL) protects the instrument and columns by removing particles and gases from solvents and other mobile phase components.
Productos recomendados
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Artículos
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.