Documentos clave
58063
Reservoir for Filtration Apparatus 1
volume 250 mL
About This Item
Código UNSPSC:
41115700
volumen
250 mL
Descripción general
Replacement glass reservoir for Supelco® mobile phase filtration apparatus 1.
Reservoir for Filtration Apparatus 1(volume 250 mL) protects the instrument and columns by removing particles and gases from solvents and other mobile phase components.
Productos recomendados
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Información legal
Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Artículos
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.