Documentos clave
Z742909
Kern FCB series bench scale
Kern FCB 6K0.5, weighing capacity 6000 g, accuracy: 0.5 g, precision: ±1.5 g
About This Item
UNSPSC Code:
41111502
NACRES:
NB.71
AC/DC input
100 - 240 V AC, universal plug set
manufacturer/tradename
Kern FCB 6K0.5
weighing capacity measuring range
6000 g, accuracy: 0.5 g, precision: ±1.5 g
General description
Bench scale with second display on the backside of the scale.
PRE-TARE function for manual subtraction of a known container weight, useful for checking fill-levels
High mobility: Battery operation option, compact construction and low weight make this balance ideal for use in several locations (production, warehouse, dispatch department, ...)
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)