Documenti fondamentali
GBL705008
Grace Bio-Labs ONCYTE® SuperNOVA nitrocellulose film slides
8-6 mm x 6 mm NC pads per slide, plain glass
Sinonimo/i:
nitrocellulose slides, protein microarrays
Scegli un formato
20 EA
CHF 728.00
CHF 728.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
20 EA
CHF 728.00
About This Item
Codice UNSPSC:
41122600
NACRES:
NB.15
CHF 728.00
Check Cart for Availability
Materiali
nitrocellulose (on glass)
Confezionamento
pkg of 20 ea
Produttore/marchio commerciale
Grace Bio-Labs 705008
Dimens. esterne
25 mm × 75 mm
Tipo di binding
high binding surface
Descrizione generale
Fluorescence detection is a preferred method for many quantitative protein microarray assaysin research and diagnostic applications due to its excellent sensitivity and potential for multiplexing. Use of multiplexed fluorescence-based assays on porous nitrocellulose though has been hampered by high backgrounds at shorter excitation wave lengths (such as the 532nm fluorescence channel). These ONCYTE™ SuperNOVA™ microporous nitrocellulose film slides increase protein microarray assay sensitivity and dynamic range using fluorescent end points at multiple wave lengths. Use of SuperNOVA™ slides can lower the limit of detection with fluorescent end points by 3 – 4 orders of magnitude over competing nitrocellulose film slides.
Note legali
NOVA is a trademark of Grace Bio-Labs, Inc.
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
SuperNOVA is a trademark of Grace Bio-Labs, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)