ETIPRQ1250-EP
Eppendorf® epT.I.P.S. standard
(dark green), size, pack of 960 ea (10 trays × 96 tips)
About This Item
Codice UNSPSC:
12352200
Prodotti consigliati
Materiali
(dark green)
Misura
L
Sterilità
non-sterile
Caratteristiche
Eppendorf Quality™
Confezionamento
pack of 960 ea (10 trays × 96 tips)
pack of 960 ea (Autoclavable refill trays)
Produttore/marchio commerciale
Eppendorf® 022494018 (NA)
Volume massimo
1.25 mL
Descrizione generale
Bulk packed in re-sealable bags.
200 μL, 300 μL and 1000 μL tips are color coded, yellow, orange and blue respectively.
200 μL, 300 μL and 1000 μL tips are color coded, yellow, orange and blue respectively.
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Quality is a trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)