Documenti fondamentali
BAF134690003
Bel-Art® Write-on Tape Dispenser
Separator Discs
About This Item
Codice UNSPSC:
55121611
NACRES:
NB.35
Confezionamento
pack of 3
Produttore/marchio commerciale
Bel-Art F13469-0003
Descrizione generale
Prevents adjacent rolls of labeling tape from binding with each other. Use when loading dispenser with more than 4 rolls of labeling tape.
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Write-on is a trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)