Documenti fondamentali
Z178365
Scienceware® dispensing jug
snap closure, prevents "glugging", pkg of 1 ea
Sinonimo/i:
5 gallon bottle, water containers, water storage
Scegli un formato
1 EA
CHF 69.50
CHF 69.50
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 69.50
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.35
CHF 69.50
Check Cart for Availability
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Bel-Art
Capacità contenitore
5 gal (20 L)
Descrizione generale
Five-gallon HDPE jug has flat sides, rounded corners, carrying handle, and bottom grip for pouring. Fill opening is 52 mm and contains a screw-cap having center thread for spigot. LDPE spigot is 1/4 turn type with 5/8 in. diameter outlet tube (1 1/4 in. long).
Jug lies on side with spigot overhanging shelf or table. Air vent, with snap closure, prevents "glugging". Spigot extends 70 mm from cap. 1 gal-molded-in graduations. 25 cm2 × 38 cm high over the handle.
Note legali
Scienceware is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)