Documenti fondamentali
WHA77073600
Whatman® 850-DS 8 channel filter plates
0.2 μm, polyethersulfone membrane, non-sterile, 50 ea
About This Item
Codice UNSPSC:
41121800
NACRES:
NC.01
Prezzi e disponibilità al momento non sono disponibili
Nome del prodotto
Whatman® 850-DS 8-channel filter plate, polyethersulfone membrane, pore size 0.2 μm, pack of 50 ea
Materiali
polyethersulfone membrane
Confezionamento
pack of 50 ea
Produttore/marchio commerciale
Whatman 770-3600
Dimensione pori
0.2 μm
Tipo di binding
non-treated surface
Descrizione generale
Save time and eliminate errors associated with manual sampling. Use Whatman 850-DS 8-channel filter plates in your Agilent 850-DS Dissolution Sampling Station
Applicazioni
The filter plates are exclusively designed for use with the optional filter module on the Agilent 850-DS Dissolution Sampling Station to simplify filter replacement between timepoints. Reliable alignment of the liquid path increases productivity in two ways. One, by reducing the risk of jamming, and two, by reducing leaks that may occur with manual sampling or other dissolution sample preparation systems.
Caratteristiche e vantaggi
  • Automated processing: up to 8 samples simultaneously
  • Readily available: in a wide range of pore sizes and materials
Note legali
Whatman is a registered trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)