Documenti
SYND452424
Synthware Dean Stark distillation receiver
20 mL, joint: ST/NS 24/40
Sinonimo/i:
Synthware D452424, Dean-Stark apparatus, Dean-Stark trap
About This Item
Codice UNSPSC:
41104812
NACRES:
NB.51
Giunto
joint: ST/NS24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware D452424
Capacità
20 mL
Descrizione generale
For easy separation of water from reaction or distillation system. Graduated with 2 mm PTFE stopcock.
Note legali
Synthware is a trademark of Kemtech America Inc.