Documenti fondamentali
DWK11-0051K
Wheaton® Microliter Snap Caps With Septa
black PTFE cap (Push fit)
Scegli un formato
100 EACH
CHF 56.90
CHF 56.90
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
100 EACH
CHF 56.90
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.11
CHF 56.90
Check Cart for Availability
Materiali
black PTFE cap (Push fit)
Caratteristiche
cap with septa
Confezionamento
pkg of 100 ea
Produttore/marchio commerciale
WHEATON® 11-0051K
Applicazioni
Chromatography
Caratteristiche e vantaggi
  • 11mm Snap Caps with installed septa offered in a wide variety of the most popular configurations.
  • Where noted, the Ring is installed between cap and septa for robotics employing magnetics to move vials.
  • Approved for GC and LC (where noted).
Note legali
WHEATON is a registered trademark of Wheaton Industries, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)