Principaux documents
TMO2403-0250
Nalgene® phthalate-free wash bottle
FEP, bottle volume 250 mL, unlabelled, case of 4 ea
Synonyme(s) :
fluid diepenser, plastic wash bottle
4 EACH
1 080.00 CHF
1 080.00 CHF
Check Cart for Availability
Changer de vue
4 EACH
1 080.00 CHF
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
1 080.00 CHF
Check Cart for Availability
Matériaux
FEP
Conditionnement
case of 4 ea
Fabricant/nom de marque
Nalgene 2403-0250
Volume du flacon
250 mL
Description générale
Ensure contamination-free dispensing with Thermo Scientific Nalgene Wash Bottles made with PTFE FEP, useful for trace element work. These phthalate-free bottles offer excellent chemical resistance and are molded entirely of fluoropolymers. Designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof† service. The precision-molded tip provides a straight, steady stream and ultimate control without sputtering.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries