Principaux documents
EP5895125000
Buckets for Eppendorf® S-4x750 Rotor
aerosol-tight capable, including plate carrier, pack of 2
2 EA
1 010.00 CHF
1 010.00 CHF
Check Cart for Availability
Changer de vue
2 EA
1 010.00 CHF
About This Item
Code UNSPSC :
41103910
Nomenclature NACRES :
NB.82
1 010.00 CHF
Check Cart for Availability
Caractéristiques
aerosol-tight capable
including plate carrier
Conditionnement
pack of 2
Fabricant/nom de marque
Eppendorf® 5895125000 (Global)
Technique(s)
centrifugation: suitable
Description générale
2 xbuckets to hold plates including plate carriers. Fits Eppendorf for Rotor S-4x750 and can be used with an aerosol-tight system.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG