Principaux documents
Z677833
Finntip® Flex pipette tips
Finntip Flex 10, sterile
960 EA
134.00 CHF
134.00 CHF
Check Cart for Availability
Changer de vue
960 EA
134.00 CHF
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.01
134.00 CHF
Check Cart for Availability
Matériaux
colorless tip
pink
Stérilité
sterile
Caractéristiques
barrier: no
Conditionnement
pack of 960 ea (10 racks of 96 tips)
Fabricant/nom de marque
Finn 94060103
Volume maximum
10 μL
Gamme de volumes
0.1-10 μL
Description générale
Finntip Flex pipette tips are easy-to-use, soft, flexible high-end tips. They are molded with diamond polished tooling for an extra smooth tip surface, which prevents liquid retention and achieves better pipetting accuracy.
Autres remarques
Browse all pipette tips
Informations légales
Finntip is a registered trademark of Thermo Fisher Scientific or its subsidiaries