Principaux documents
SYNA284250
Synthware anti-splash adapter
250 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 24/40
Synonyme(s) :
Synthware A284250, rotary evaporator bump trap, bump-trap
1 EA
123.00 CHF
123.00 CHF
Check Cart for Availability
Changer de vue
1 EA
123.00 CHF
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
123.00 CHF
Check Cart for Availability
Joint
bottom joint: ST/NS24/40
joint: ST/NS top24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A284250
Capacité
250 mL
Catégories apparentées
Description générale
This design prevents possible contamination from rotary evaporator.
Informations légales
Synthware is a trademark of Kemtech America Inc.