Principaux documents
SYNA282924
Synthware anti-splash adapter
100 mL, top joint: ST/NS 29/42, bottom joint: ST/NS 24/40
Synonyme(s) :
Synthware A282924, rotary evaporator adapter
1 EA
122.00 CHF
122.00 CHF
Check Cart for Availability
Changer de vue
1 EA
122.00 CHF
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
122.00 CHF
Check Cart for Availability
Joint
bottom joint: ST/NS24/40
joint: ST/NS top29/42
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A282924
Capacité
100 mL
Catégories apparentées
Description générale
This design prevents possible contamination from rotary evaporator.
Informations légales
Synthware is a trademark of Kemtech America Inc.