Principaux documents
Z684511
Ace rotary evaporator bump trap
1 EA
153.00 CHF
153.00 CHF
Check Cart for Availability
Changer de vue
1 EA
153.00 CHF
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
153.00 CHF
Check Cart for Availability
Questions
  1. What are the sizes of the tapered joint and the cone? These are not on the product description at time of asking.
    1 answer
    1. The product description of Z684511 is: Trap, bump, elliptical, 24/40 top, 24/40 bottom