Merck
HPLC Analysis of Norvaline Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Norvaline Enantiomers on Astec® CHIROBIOTIC® T application for HPLC
Materialien
Analytische Säule
Produkt-Nr.
Beschreibung
Preisangaben
Astec® CHIROBIOTIC® T Chirale HPLC-Säule
5 μm particle size, L × I.D. 25 cm × 4.6 mm
Standard
Produkt-Nr.
Beschreibung
Preisangaben
DL-Norvalin
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] water; [B] ethanol; (90:10, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 210 nm
injection
5 μL
sample
5 mg/mL in methanol