Amyloid β-protein fragment 10-35
Materialien
Analytische Säule
Produkt-Nr.
Beschreibung
Preisangaben
CONDITIONS
column
Discovery BIO Wide Pore C18, 25 cm × 4.6 mm I.D., 5 μm particles (568223-U)
mobile phase
[A] 0.1% TFA in water; [B] 0.1% TFA in water:0.1% TFA in acetonitrile; (25:75)
flow rate
1.3 mL/min
column temp.
30 °C
detector
UV, 215 nm
injection
10 μL, 1 mg/mL in water