Merck
V900789
Sigma-Aldrich
3-Hydroxy-benzoesäure
Vetec, reagent grade, 98%
About This Item
Lineare Formel:
HOC6H4CO2H
CAS-Nummer:
Molekulargewicht:
138.12
Beilstein:
508160
EG-Nummer:
MDL-Nummer:
UNSPSC-Code:
12352100
PubChem Substanz-ID:
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Qualität
reagent grade
Produktlinie
Vetec
Assay
98%
mp (Schmelzpunkt)
200-203 °C (lit.)
SMILES String
OC(=O)c1cccc(O)c1
InChI
1S/C7H6O3/c8-6-3-1-2-5(4-6)7(9)10/h1-4,8H,(H,9,10)
InChIKey
IJFXRHURBJZNAO-UHFFFAOYSA-N
Rechtliche Hinweise
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Piktogramme
Exclamation mark
Signalwort
Warning
Gefahreneinstufungen
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Lagerklassenschlüssel
11 - Combustible Solids
WGK
WGK 3
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Fredrik L Nordström et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 28(5), 377-384 (2006-06-20)
Solution and solid-state properties of m-hydroxybenzoic acid have been investigated. Two polymorphs were found where the monoclinic modification exhibits a higher stability than the orthorhombic form. The solubility of the monoclinic polymorph was determined between 10 and 50 degrees C
Ya-Wen He et al.
Molecular plant-microbe interactions : MPMI, 24(8), 948-957 (2011-05-05)
Xanthomonas campestris pv. campestris produces a membrane-bound yellow pigment called xanthomonadin. A diffusible factor (DF) has been reported to regulate xanthomonadin biosynthesis. In this study, DF was purified from bacterial culture supernatants using a combination of solvent extraction, flash chromatography
Yi-Li Lin et al.
Journal of hazardous materials, 137(1), 324-331 (2006-04-11)
This research is dedicated to determine the rejection ratios of disinfection by-products (DBPs) precursors including resorcinol, phloroglucinol, 3-hydroxybenzoic acid, and tannic acid solution in the presence of calcium by nanofiltration with NF70 membrane. The rejections of these model compounds also
Xianli Wu et al.
Molecular nutrition & food research, 53 Suppl 1, S76-S84 (2009-02-10)
Black raspberries (BRB) contain high levels of polyphenols and have been demonstrated to be chemopreventive. In order to investigate the underlying mechanism and study the metabolism of anthocyanins, pigs were fed freeze-dried BRB powder or purified diet (control) and three
N Laman et al.
Applied spectroscopy, 62(3), 319-326 (2008-03-15)
Low frequency vibrational modes of pharmaceutical molecules are dependent on the molecule as a whole and can be used for identification purposes. However, conventional Fourier transform far-infrared spectroscopy (FT-IR) and terahertz time-domain spectroscopy (THz-TDS) often result in broad, overlapping features