Merck
Z760986
PCV cell counting tubes
capillary graduations only, no cap
Synonym(e):
PCV Tubes
Größe auswählen
50 EA
CHF 229.00
CHF 229.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
50 EA
CHF 229.00
About This Item
UNSPSC-Code:
41121706
NACRES:
NB.22
CHF 229.00
Check Cart for Availability
Materialien
(PCV)
colorless tube
conical bottom
straight neck
Sterilität
non-sterile
Verpackung
pack of 50 ea
Hersteller/Markenname
TPP 87005
Verfügbarkeit
not available in (available only in North America)
Allgemeine Beschreibung
The packed cell volume (PCV) tube is a quick and reproducible measuring technique of the cell volume that is suitable to monitor cell growth (cell mass).
Leistungsmerkmale und Vorteile
  • Present with or without scale.
  • Fits in microcentrifuges.
  • Caps available if required.
  • Ideal to monitor cell growth
Empfehlung
Produkt-Nr.
Beschreibung
Preisangaben
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Yan Xu et al.
Journal of nanobiotechnology, 15(1), 50-50 (2017-07-12)
Increasing production of nanomaterials requires fast and proper assessment of its potential toxicity. Therefore, there is a need to develop new assays that can be performed in vitro, be cost effective, and allow faster screening of engineered nanomaterials (ENMs). Herein
Xiqian Jiang et al.
ACS chemical biology, 10(3), 864-874 (2014-12-23)
Glutathione (GSH) plays an important role in maintaining redox homeostasis inside cells. Currently, there are no methods available to quantitatively assess the GSH concentration in live cells. Live cell fluorescence imaging revolutionized the field of cell biology and has become