Merck
Z740356
Nunc® petri dishes
H 150 mm × 20 mm, surface area × working volume 145 cm2 × 35 mL, lid
Synonym(e):
culture dishes, disposable petri dishes, plastic petri dishes
Größe auswählen
80 EA
CHF 230.00
CHF 230.00
Voraussichtliches Versanddatum28. Mai 2025
Größe auswählen
Ansicht ändern
80 EA
CHF 230.00
About This Item
UNSPSC-Code:
41121812
NACRES:
NB.14
CHF 230.00
Voraussichtliches Versanddatum28. Mai 2025
Materialien
clear polystyrene
Sterilität
sterile
Leistungsmerkmale
lid
Verpackung
pack of 80 ea
Hersteller/Markenname
Nunc 168381
Höhe
150 mm × 20 mm
Schalendurchm. × H
150 mm × 20 mm
surface area × working volume
145 cm2 × 35 mL
Bindungsart
Nunclon Delta surface
Allgemeine Beschreibung
Polystyrene provides high optical quality-especially when microscopic examination is required.
Includes:
  • A certificate which assures user of biological performance as well as traceability
  • Top and bottom stacking rings for easier handling and more stable stacking
  • Tested for cell growth and plating efficiency
  • Radiation sterilized
  • Air vents provide gas exchange
  • For cloning experiments and plating efficiency determinations
  • With lid
Rechtliche Hinweise
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)