Merck
Z740290
RotoFlex® Plus bench top tube rotator
AC/DC input 220 - 240 V AC (Europlug flat 2-pin & UK 3-pin plug)
Synonym(e):
Western Blot hybridization, sample mixing, tube rotator
About This Item
UNSPSC-Code:
41103811
NACRES:
NB.73
Leistungsmerkmale
unit is supplied w/rotisseries, 1 pack of each size
Hersteller/Markenname
Cole Parmer 439741
Allgemeine Beschreibung
The RotoFlex Plus is a variable speed, digitally controlled tube rotator; accommodating tubes from 0.5 - 50 mL by using included rotisserie panels.
The easy to operate key pad allows for speed adjustments between 10 - 40 rpm. In addition to the variable speed feature the unit can also be operated in an oscillating mode. In oscillation mode the rotisseries rotate 40°, pause, then oscillate back and forth five times before rotating another 40° to repeat the process. After oscillating for 360°, the rotisserie rotates a complete turn before reverting back to the oscillation cycle.
The RotoFlex Plus can also be used with a magnetic platform (sold separately) to optimize Western Blot hybridization. Simply attach your plastic bag containing membrane & antibody solution to the plate using the metal strips provided.
Features:
To change the rotation direction - simply touch the rotisserie paddle while in motion and the RotoFlex Plus automatically begins to rotate in the opposite direction. The RotoFlex Plus can be used in incubators & cold room settings. Unit is supplied with base unit and rotisseries (1 pack of each size).
Ideal for:
  • Hybridization
  • DNA Purification
  • Sample mixing/resuspension
  • Protein Binding
  • Immunoprecipitation
Rechtliche Hinweise
RotoFlex is a registered trademark of Argos Technologies, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)