Merck
Z740157
LifeSep magnetic separation stand
50SX, for 50 mL tubes
Synonym(e):
magnetic separation equipment
Größe auswählen
1 EA
CHF 959.00
CHF 959.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 959.00
About This Item
UNSPSC-Code:
41100000
NACRES:
NA.56
CHF 959.00
Check Cart for Availability
Leistungsmerkmale
for 50 mL tubes
Hersteller/Markenname
Dexter Magnetic Technologies 2501022-1
Verwandte Kategorien
Allgemeine Beschreibung
Magnetic separators for single tubes are durable and easy-to-use. For use in cell sorting, RNA and DNA isolation and purification of biomolecules. Fast and easy aspiration is achieved with our Neodymium-Iron-Boron (Nd-Fe-B) magnet configuration.
  • Patented Nd-Fe-B magnet configuration for optimal results
  • Open-faced design for clear visibility
  • Rapid separation of micro- and nanometer beads
  • Easy aspiration
  • Superior particle retention
  • Removable base for easy cleaning
  • Durable and easy to use
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)