Merck
Z601527
Hettich® centrifuge rotors
for UNIVERSAL 320/320R Centrifuge, Swing-out Rotor, 4 place
Größe auswählen
1 EA
CHF 418.00
CHF 418.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 418.00
About This Item
UNSPSC-Code:
41103900
NACRES:
NB.78
CHF 418.00
Check Cart for Availability
Leistungsmerkmale
to hold 4 x Suspensions (Models 1345, 1346, 1366, 1369, 1370, 1372, 1741, 1742, 1745, 1746)
Verpackung
pack of 1 ea
Hersteller/Markenname
Hettich 1624
Parameter
2,665 max. RCF
Methode(n)
centrifugation: suitable
Kompatibilität
for use with centrifuge models Universal 320/320R and Rotofix 32A
Allgemeine Beschreibung
The 4-place rotor 1624 (Z601527) is best suited for laboratories not exclusively involved with cytological examinations. Both tubes and cytological prepartions can be centrifuged without requiring a rotor change. Changing the buckets/carriers is all that is needed. Rotor 1624 (Z601527) accommodates cyto bucket/carriers 1660 and 1680.
Rechtliche Hinweise
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)